Läs mer
Caspofungin Acetate, TRC
Produktkod. 30003329
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30003329 5 mg
30003433 10 mg
30010003 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30003329 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1849005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 179463-17-3
Synonym Pneumocandin B0, 1-[(4R,5S)-5-[(2-aminoethyl)amino]-N2-[(10R,12S)-10,12-dimethyl-1-oxotetradecyl]-4-hydroxy-L-ornithine]-5-[(3R)-3-hydroxy-L-ornithine]-, acetate (1:2)∣Pneumocandin B0, 1-[(4R,5S)-5-[(2-aminoethyl)amino]-N2-[(10R,12S)-10,12-dimethyl-1-oxotetradecyl]-4-hydroxy-L-ornithine]-5-(threo-3-hydroxy-L-ornithine)-, diacetate (salt)∣Pneumocandin B0, 1-[(4R,5S)-5-[(2-aminoethyl)amino]-N2-[(10R,12S)-10,12-dimethyl-1-oxotetradecyl]-4-hydroxy-L-ornithine]-5-[(3R)-3-hydroxy-L-ornithine]-, diacetate (salt) (9CI)∣Cancidas∣Caspofungin acetate∣Caspofungin diacetate∣L 743872∣L 743873∣MK 0991
Specifikationer
Renhet >95% (HPLC)
Formatera Neat
Molekylvikt (g/mol) 1213.42
Produkttyp API