Läs mer
Caspofungin Acetate, TRC
Produktkod. 30010003
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010003 50 mg
30003329 5 mg
30003433 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010003 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC18490050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 179463-17-3
Synonym Pneumocandin B0, 1-[(4R,5S)-5-[(2-aminoethyl)amino]-N2-[(10R,12S)-10,12-dimethyl-1-oxotetradecyl]-4-hydroxy-L-ornithine]-5-[(3R)-3-hydroxy-L-ornithine]-, acetate (1:2)∣Pneumocandin B0, 1-[(4R,5S)-5-[(2-aminoethyl)amino]-N2-[(10R,12S)-10,12-dimethyl-1-oxotetradecyl]-4-hydroxy-L-ornithine]-5-(threo-3-hydroxy-L-ornithine)-, diacetate (salt)∣Pneumocandin B0, 1-[(4R,5S)-5-[(2-aminoethyl)amino]-N2-[(10R,12S)-10,12-dimethyl-1-oxotetradecyl]-4-hydroxy-L-ornithine]-5-[(3R)-3-hydroxy-L-ornithine]-, diacetate (salt) (9CI)∣Cancidas∣Caspofungin acetate∣Caspofungin diacetate∣L 743872∣L 743873∣MK 0991
Specifikationer
Renhet >95% (HPLC)
Formatera Neat
Molekylformel C56H96N10O19
MDL-nummer MFCD08141839
Molekylvikt (g/mol) 1213.42, 1213.44
InChI-nyckel OGUJBRYAAJYXQP-LLXMLGLCSA-N
LEDER CC(O)=O.CC(O)=O.[H][C@@]12C[C@@H](O)CN1C(=O)[C@@]([H])(NC(=O)[C@H](C[C@@H](O)[C@@H](NCCN)NC(=O)[C@]1([H])[C@@H](O)CCN1C(=O)[C@@]([H])(NC(=O)[C@@]([H])(NC2=O)[C@H](O)[C@@H](O)C1=CC=C(O)C=C1)[C@H](O)CCN)NC(=O)CCCCCCCCC(C)CC(C)CC)[C@@H](C)O
IUPAC-namn N-[(3S,6S,9S,11R,15S,18S,20R,21S,24S,25S)-3-[(1R)-3-amino-1-hydroxypropyl]-21-[(2-aminoethyl)amino]-6-[(1S,2S)-1,2-dihydroxy-2-(4-hydroxyphenyl)ethyl]-11,20,25-trihydroxy-15-[(1R)-1-hydroxyethyl]-2,5,8,14,17,23-hexaoxo-1,4,7,13,16,22-hexaazatricyclo[22.3.0.0⁹,¹³]heptacosan-18-yl]-10,12-dimethyltetradecanamide; bis(acetic acid)
Produkttyp API