Läs mer
Fisherbrand™ Catalyst PharMed™ BPT Formulation Stopped Tubing Assembly
Produktkod. 15581325 Handla allt Fisherbrand Produkter
Diameter (metrisk) Inre:
0.8 mm
1.6 mm
3.2 mm
4.8 mm
6.4 mm
8 mm
9.5 mm
Tjocklek (metrisk):
1.6 mm
2.4 mm
Förpackningsstorlek:
Paket med 12
Produktkod. 15581325
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Preconfigured ready-to-use links and assemblies.
  • Available in Platinum-Cured Silicone, TPEs, and PVC
  • Ensures optimal amount of tubing is in pump head
Specifikationer
Beskrivning Stopped Tubing Assembly
Diameter (metrisk) Inre 6.4 mm
Tjocklek (metrisk) 2.4 mm