Learn More
Cole-Parmer™ Catalyst Pump Tubing
Designed especially for handling food products. Cole-Parmer™ Catalyst Tygon E-Food Pump Tubing is compatible with Catalyst FH100, FH100D Pumps.
Brand: Cole-Parmer™ MFLX97000-72
Additional Details : Weight : 0.63000kg
Description
Designed for use with Catalyst FH100, FH100D, FH100X and FH100DX Pumps. Cole-Parmer™ Catalyst Pump Tubing is available four formulations in 15 meter lengths.
- Transparent pump tubing
- For Catalyst FH100, FH100D, FH100X and FH100DX Pumps
- Four formulations available to cover most chemical compatible applications
- To help simplify choices, a platinum-cured silicone, two TPEs and a long life PVC tubing material are offered.
- Standard 15 meter lengths
- Available in both thin and thick wall configurations
- FH100 and FH100D utilize tubing with a wall thickness of 1.6mm
- FH100X and FH100DX utilize tubing with a wall thickness of 2.4mm
Specifications
Catalyst Pump Coiled Tubing | |
3.2 mm | |
49.2 ft. | |
0.094 in. | |
Catalyst Pump models: FH100, FH100D, FH100X and FH100DX |
0.12 in. | |
Silicone (Platinum-Cured) | |
1500 cm | |
1.6 mm | |
Clear |