Läs mer
Catapol, TRC
Produktkod. 30363365
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30363365 100 mg
1 options
Produktkod. 30363365 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC215150100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Catalpol
CAS 2415-24-9
Anteckningar om renhetsgrad HPLC
Molekylformel C15 H22 O10
Kvantitet 100 mg
Synonym β-D-Glucopyranoside, (1aS,1bS,2S,5aR,6S,6aS)-1a,1b,2,5a,6,6a-hexahydro-6-hydroxy-1a-(hydroxymethyl)oxireno[4,5]cyclopenta[1,2-c]pyran-2-yl, Catalpol (7CI,8CI), β-D-Glucopyranoside, 1a,1b,2,5a,6,6a-hexahydro-6-hydroxy-1a-(hydroxymethyl)oxireno[4,5]cyclopenta[1,2-c]pyran-2-yl, [1aS-(1aα,1bβ,2β,5aβ,6β,6aα)]-, (1aS,1bS,2S,5aR,6S,6aS)-1a,1b,2,5a,6,6a-Hexahydro-6-hydroxy-1a-(hydroxymethyl)oxireno[4,5]cyclopenta[1,2-c]pyran-2-yl β-D-glucopyranoside, 7,8-Epoxyaucubin, Aucubin 7,8-epoxide, Catalpinoside, Catapol, De(p-hydroxybenzoyl)catalposide
InChI formel InChI=1S/C15H22O10/c16-3-6-9(19)10(20)11(21)14(23-6)24-13-7-5(1-2-22-13)8(18)12-15(7,4-17)25-12/h1-2,5-14,16-21H,3-4H2/t5-,6-,7-,8+,9-,10+,11-,12+,13+,14+,15-/m1/s1
LEDER OC[C@H]1O[C@@H](O[C@@H]2OC=C[C@H]3[C@H](O)[C@@H]4O[C@]4(CO)[C@@H]23)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC-namn (2S,3R,4S,5S,6R)-2-[[(1aS,1bS,2S,5aR,6S,6aS)-6-hydroxy-1a-(hydroxymethyl)-2,5a,6,6a-tetrahydro-1bH-oxireno[5,6]cyclopenta[1,3-c]pyran-2-yl]oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Molekylvikt (g/mol) 362.33
Formel vikt 362.1213