Läs mer
(+/-)-Catechin xHydrate, TRC
Produktkod. 30134914
Change view
Kvantitet:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134914 1 g
30132593 500 mg
30007796 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134914 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC2175201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7295-85-4
Molekylvikt (g/mol) 290.27
IUPAC-namn (2R,3S)-2-(3,4-dihydroxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol
LEDER O[C@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3ccc(O)c(O)c3
Specifikationer
Kemiskt namn eller material DL-Catechin
Molekylformel C15 H14 O6
Kvantitet 1 g
Synonym 2H-1-Benzopyran-3,5,7-triol, 2-(3,4-dihydroxyphenyl)-3,4-dihydro-, trans-(±)-, Catechol, (±)- (8CI), rel-(2R,3S)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-3,5,7-triol, (±)-Catechin, (±)-Catechol, 2H-1-Benzopyran-3,5,7-triol, 2-(3,4-dihydroxyphenyl)-3,4-dihydro-, trans-, DL-Catechin, Racemic catechin, dl-Catechin, dl-Catechol, rac-Catechin
InChI formel InChI=1S/C15H14O6/c16-8-4-11(18)9-6-13(20)15(21-14(9)5-8)7-1-2-10(17)12(19)3-7/h1-5,13,15-20H,6H2/t13-,15+/m0/s1
Formel vikt 290.079