Läs mer
Catechol-13C6, TRC
Produktkod. 30010356
Change view
Kvantitet:
1 mg
2.5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30010356 10 mg -
30008457 1 mg -
30009318 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010356 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC21517710MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 115881-16-8
Molekylformel 13C6 H6 O2
Molekylvikt (g/mol) 116.07
Synonym 1,2-Benzenediol-1,2,3,4,5,6-13C6 (9CI), Catechol-1,2,3,4,5,6-13C6, Catechol 13C6, Benzene-1,2-diol 13C6
IUPAC-namn (1,2,3,4,5,6-^{13}C_{6})cyclohexa-2,4,6-triene-1,2-diol
LEDER O[13c]1[13cH][13cH][13cH][13cH][13c]1O
Specifikationer
Kemiskt namn eller material Catechol 13C6
Kvantitet 10 mg
InChI formel InChI=1S/C6H6O2/c7-5-3-1-2-4-6(5)8/h1-4,7-8H/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vikt 116.0569