Läs mer
Catechol-O-sulphate Potassium Salt, TRC
Produktkod. 30132888
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132888 250 mg
17921795 25 mg
17903245 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132888 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS336750250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 68944-25-2
Molekylformel C6H6O5SK
Molekylvikt (g/mol) 228.26
Synonym 2-Hydroxyphenyl Hydrogen Sulfate Ester Potassium Salt
IUPAC-namn potassium;(2-hydroxyphenyl) sulfate
LEDER [K+].Oc1ccccc1OS(=O)(=O)[O-]
Specifikationer
Kemiskt namn eller material Catechol-O-sulphate Potassium Salt
Kvantitet 250 mg
InChI formel InChI=1S/C6H6O5S.K/c7-5-3-1-2-4-6(5)11-12(8,9)10;/h1-4,7H,(H,8,9,10);/q;+1/p-1
Formel vikt 227.9495