missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ β-Catenin Rabbit Monoclonal Antibody

Ensure accurate and reproducible results in immunohistochemistry applications with this antibody, part of a comprehensive antibody portfolio.

Brand:  Epredia™ RM-2101-RQ

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 12693597

  • 7560.00 SEK / 12mL
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The catenins (α, β and γ) are ubiquitously expressed cytoplasmic proteins associated with E-cadherin at cellular junctions. β-catenin also binds to N-cadherin and co-immunoprecipitates with APC. Cadherin/catenin complexes are linked to the cytoskeleton via a direct association between α-actinin and α-catenin. Increased tyrosine phosphorylation can disrupt catenin-cadherin complexes, influencing cellular adhesion. Mutations in the catenin gene, CTNNB1, have been implicated in its accumulation and various forms of carcinomas.

Host Species: Rabbit

Clone: E247

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide derived from near the N-terminus of human β-catenin

Molecular Weight: 92 kDa

Positive Control: Breast CA

Cellular Localization: Cell Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Catenin, beta
Monoclonal
Unconjugated
A synthetic peptide derived from near the N-terminus of human beta-catenin
IVD
Primary
Immunohistochemistry (Paraffin)
E247
Rabbit
12 mL
Cancer and Tumor Biology
Ready-to-Use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers