missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Cathepsin B Recombinant Rabbit Monoclonal Antibody (3X1D6)

Produktkod. 30131330
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30131330 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30131330 Leverantör Invitrogen™ Leverantörsnummer MA555403

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Recombinant Monoclonal Antibody

Immunogen Sequence: CCLLVLANAR SRPSFHPLSD ELVNYVNKRN TTWQAGHNFY NVDMSYLKRL CGTFLGGPKP PQRVMFTEDL KLPASFDARE QWPQCPTIKE IRDQGSCGSC WAFGAVEAIS DRICIHTNAH VSVEVSAEDL LTCCGSMCGD GCNGGYPAEA WNFWTRKGLV SGGLYESHVG CRPYSIPPCE HHVNGSRPPC TGEGDTPKCS KICEPGYSPT YKQDKHYGYN SYSVSNSEKD IMAEIYKNGP VEGAFSVYSD FLLYKSGVYQ HVTGEMMGGH AIRILGWGVE NGTPYWLVAN SWNTDWGDNG FFKILRGQDH CGIESEVVAG IPRTDQYWEK I.

Cathepsin B is a gene located on chromosome 8p23.1 that encodes a lysosomal cysteine protease involved in intracellular protein degradation. As a member of the cathepsin family, Cathepsin B plays a crucial role in the lysosomal pathway, facilitating the breakdown of proteins into amino acids, which can then be recycled for cellular functions. In addition to its proteolytic activity within lysosomes, Cathepsin B is also implicated in extracellular matrix remodeling, cellular invasion, and apoptotic pathways. Its expression and activity are often upregulated in various cancer types, including breast, colon, and pancreatic cancers, where it contributes to tumor progression and metastasis by degrading extracellular matrix components and promoting tumor cell invasion. Due to these roles, Cathepsin B is considered a potential biomarker for cancer prognosis and a target for therapeutic intervention aimed at inhibiting cancer cell dissemination. Research continues to explore its broader implications in diseases related to protease activity and inflammation.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Cathepsin B
Användningsområden ELISA, Western Blot, Immunocytochemistry
Klassificering Recombinant Monoclonal
Klona 3X1D6
Koncentration 0.6 mg/mL
Konjugera Unconjugated
Formulering PBS with 50% glycerol, 0.05% BSA and 0.05% ProClin 300; pH 7.3
Gen Ctsb
Genaccessionsnr. P07858
Gene Alias amyloid precursor protein secretase; APP secretase; APPS; cathepsin B; Cathepsin B heavy chain; Cathepsin B light chain; Cathepsin B1; CB; CPSB; Ctsb; cysteine protease; RSG-2
Gensymboler Ctsb
Värd art Rabbit
Immunogen Recombinant protein (or fragment).
Reningsmetod Protein A
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 1508
Målarter Human
Innehåll och lagring -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.