missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin L Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10938-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a lysosomal cysteine proteinase that plays a major role in intracellular protein catabolism. Its substrates include collagen and elastin, as well as alpha-1 protease inhibitor, a major controlling element of neutrophil elastase activity. The encoded protein has been implicated in several pathologic processes, including myofibril necrosis in myopathies and in myocardial ischemia, and in the renal tubular response to proteinuria. This protein, which is a member of the peptidase C1 family, is a dimer composed of disulfide-linked heavy and light chains, both produced from a single protein precursor. At least two transcript variants encoding the same protein have been found for this gene.Specifications
Cathepsin L | |
Polyclonal | |
Unconjugated | |
CTSL | |
cathepsin L1, CATL, CTSL, CTSL1, Major excreted protein, MEP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1514 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P07711 | |
CTSL | |
Cathepsin L Fusion Protein Ag1373 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |