missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CATSPERB Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CATSPERB |
---|---|
Concentration | 0.4 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16825045
|
Proteintech
26633-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16815045
|
Proteintech
26633-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CATSPERB, also named as C14orf161 and CatSper-beta, is involved in sperm cell hyperactivation via its association with CATSPER1. CATSPERB has two isoforms with MW 127 kDa and 72 kDa.Specifications
CATSPERB | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9H7T0 | |
79820 | |
CATSPERB Fusion Protein Ag23393 | |
Primary | |
-20°C | |
CATSPERB |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C14orf161, CatSper beta, CatSper(beta), CATSPERB | |
CATSPERB | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |