missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ CACNA1I Polyclonal Antibody
GREENER_CHOICE

Produktkod. 11566052
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
Förpackningsstorlek:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11566052 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11566052 Leverantör Invitrogen™ Leverantörsnummer PA112666

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

A suggested positive control for this product is rat brain lysate.

CaV3.3 encodes the pore-forming alpha subunit of a voltage gated calcium channel. The encoded protein is a member of a subfamily of calcium channels referred to as is a low voltage-activated, T-type, calcium channel. The channel encoded by CaV3.3 is characterized by a slower activation and inactivation compared to other T-type calcium channels. CaV3.3 may be involved in calcium signaling in neurons. Alternate splicing results in multiple transcript variants of CaV3.3. Voltage-gated calcium channels (CaV) are present in the membrane of most excitable cells and mediate calcium influx in response to depolarization, an proteins such as CaV3.3 regulate intracellular processes such as contraction, secretion, neurotransmission and gene expression. Diseases associated with CACNA1I include Childhood Absence Epilepsy.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen CACNA1I
Användningsområden Western Blot
Klassificering Polyclonal
Koncentration 1 mg/mL
Konjugera Unconjugated
Formulering PBS with 0.08% sodium azide
Gen CACNA1I
Genaccessionsnr. Q9P0X4
Gene Alias Ca(v)3.3; CACNA1I; calcium channel, voltage-dependent, alpha 1I subunit; calcium channel, voltage-dependent, T type, alpha 1I subunit; calcium voltage-gated channel subunit alpha1 I; Cav3.3; CaVT.3; hypothetical gene supported by NM_020084; KIAA1120; low voltage-activated T-type calcium channel alpha-1 subunit (CACNA1I); low-voltage-activated calcium channel alpha13.3; RP1-172B20.4; voltage-dependent calcium channel; voltage-dependent T-type calcium channel subunit alpha-1I; Voltage-gated calcium channel subunit alpha Cav3.3
Gensymboler CACNA1I
Värd art Rabbit
Immunogen Synthetic peptide, conjugated to KLH, corresponding to the rat alpha1I calcium channel.
Reningsmetod Ammonium sulfate precipitation
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 8911
Målarter Human
Innehåll och lagring -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.