Läs mer
STEMCELL Technologies CAY10585
Beskrivning
- CAY10585 inhibits hypoxia inducible factor 1 (HIF-1) accumulation in a concentration and time-dependent manner and significantly suppresses transcriptional activity of HIF-1 target genes (Lee et al.)
- Inhibits BMP9- and HIF-1-induced osteogenic differentiation in mesenchymal stem cells (Hu et al.)
- Rescues cardiomyocyte differentiation in Fgfr1(-/-) mouse embryonic stem cells (Crescini et al.)
Specifikationer
Specifikationer
| CAS | 934593-90-5 |
| Synonym | HIF-1α inhibitor; Hypoxia Inducible Factor-1α Inhibitor |
| Kvantitet | 10 mg |
| Molekylformel | C26H29NO5 |
| Renhet | ≥97% |