Läs mer
CB30865, MedChemExpress
Produktkod. 16488647
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16488647 5 mg
16426048 10 mg
16422187 50 mg
16406456 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16488647 Leverantör Medchem Express Leverantörsnummer HY14373
MedChemExpress CB30865 (ZM 242421) is a nicotinamide phosphoribosyltransferase (Nampt) inhibitor, with potent cytotoxicity. CB30865 is highly potent against a variety of human tumour cell lines (IC50 values in the 1-10 nM range).
CB30865 (ZM 242421) is a nicotinamide phosphoribosyltransferase (Nampt) inhibitor, with potent cytotoxicity. CB30865 is highly potent against a variety of human tumour cell lines (IC50 values in the 1-10 nM range).
Specifikationer
Kemiskt namn eller material CB30865
CAS 206275-15-2
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C26H22BrN5O2
Kvantitet 5 mg
Synonym ZM 242421
Löslighetsinformation DMSO : 33.33 mg/mL (64.54 mM; Need ultrasonic)
LEDER O=C(NCC1=CC=CN=C1)C2=CC=C(N(CC3=CC4=C(N=C(C)NC4=O)C=C3Br)CC#C)C=C2
Molekylvikt (g/mol) 516.39
Formel vikt 516.39
Procent renhet 98.05%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death