Läs mer
Tocris Bioscience™ CBM 301940
Beskrivning
CBM 301940 is a potent malonyl-CoA decarboxylase (MCD) inhibitor (IC50 = 23 nM). Suppresses malonyl-CoA degradation and fatty acid oxidation in isolated rat hearts, leading to secondary increase in glucose oxidation. Improves cardiac function during reperfusion of ischemic isolated rat hearts. Orally bioavailable.
Specifikationer
Specifikationer
| Kemiskt namn eller material | tert-Butyl 3-(5-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)-4,5-dihydroisoxazole-3-carboxamido)butanoate |
| CAS | 902146-11-6 |
| Kvantitet | 10 mg |
| Molekylformel | C15H20F6N2O5 |
| Formel vikt | 422.32 |
| Renhet | 98% |