missing translation for 'onlineSavingsMsg'
Learn More

CBY1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12239-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16869833

  • 4721.67 SEK / 150µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Beta-catenin is a transcriptional activator and oncoprotein involved in the development of several cancers. The protein encoded by this gene interacts directly with the C-terminal region of beta-catenin, inhibiting oncogenic beta-catenin-mediated transcriptional activation by competing with transcription factors for binding to beta-catenin. Two transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

CBY1
Polyclonal
Unconjugated
CBY1
arb1, ARPP binding protein, C22orf2, CBY, CBY1, chibby homolog 1 (Drosophila), Cytosolic leucine rich protein, HS508I15A, PGEA1, PIGEA 14, PIGEA14, Protein chibby homolog 1
Rabbit
Antigen Affinity Chromatography
RUO
246768, 25776, 73739
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8K4I6, Q9D1C2, Q9Y3M2
Cby1
CBY1 Fusion Protein Ag2882
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers