missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCBL1/KAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12156-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Kynurenine aminotransferases KAT I, KAT II, and KAT III belong to the class-I pyridoxal-phosphate-dependent aminotransferase family. KAT I is a cytoplasmic protein involved in glutamine catabolism. KAT I functions in the catalysis of the transamination of L-kinurenine to form kynurenic acid, a neuroprotective and anticonvulsant metabolite of tryptophan. Kynurenic acid is involved in synaptic transmission and has been implicated in a number of neurological disorders including schizophrenia and Huntington′s disease. KAT I also functions in the metabolism of cysteine conjugates in some halogenated alkenes and alkanes to form reactive metabolites. KAT I has three isoforms. Isoform 1is the full length protein, isoform 2 lacks amino acids 68-117 and isoform 3 lacks amino acids 251-422. Based on sequence similarity, KAT I is thought to function as a homodimer.Specifications
CCBL1/KAT1 | |
Polyclonal | |
Unconjugated | |
KYAT1 | |
CCBL1, Glutamine transaminase K, GTK, KAT1, KATI, Kynurenine aminotransferase I | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
311844, 70266, 883 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q08415, Q16773, Q8BTY1 | |
Kyat1 | |
CCBL1/KAT1 Fusion Protein Ag2828 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |