missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC109B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20387-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Negatively regulates the activity of MCU, the mitochondrial inner membrane calcium uniporter, and thereby modulates calcium uptake into the mitochondrion. Does not form functional calcium channels by itself. Mitochondrial calcium homeostasis plays key roles in cellular physiology and regulates cell bioenergetics, cytoplasmic calcium signals and activation of cell death pathways.Specifications
CCDC109B | |
Polyclonal | |
Unconjugated | |
MCUB | |
CCDC109B, MCUb | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55013 | |
-20°C | |
Liquid |
Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NWR8 | |
MCUB | |
CCDC109B Fusion Protein Ag14225 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |