missing translation for 'onlineSavingsMsg'
Learn More

CCDC137 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 27201-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16825475

  • 1656.73 SEK / 20µL
Estimated Shipment: 22-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CCDC137 is also known as the retinoic acid resistance factor protein.
Specifications

Specifications

CCDC137
Polyclonal
Unconjugated
CCDC137
CCDC137
Rabbit
Antigen Affinity Chromatography
RUO
339230
-20°C
Liquid
Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6PK04
CCDC137
CCDC137 Fusion Protein Ag23942
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers