missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC137 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27201-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
CCDC137 is also known as the retinoic acid resistance factor protein.Specifications
CCDC137 | |
Polyclonal | |
Unconjugated | |
CCDC137 | |
CCDC137 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
339230 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6PK04 | |
CCDC137 | |
CCDC137 Fusion Protein Ag23942 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |