missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC21 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26314-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
All isoforms of CCDC21 can be detected by this antibody.
This gene encodes a protein that belongs to the centrosome-associated family of proteins. The centrosome is a subcellular organelle in the animal cell that functions as a microtubule organizing center and is involved in cell-cycle progression. Alternate splicing results in multiple transcript variants.Specifications
CCDC21 | |
Polyclonal | |
Unconjugated | |
CEP85 | |
CCDC21, CEP85 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64793 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6P2H3 | |
CEP85 | |
CCDC21 Fusion Protein Ag23636 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |