missing translation for 'onlineSavingsMsg'
Learn More

CCDC27 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 23769-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16882435

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The function of this protein remains unknown.
Specifications

Specifications

CCDC27
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q2M243
CCDC27
Rabbit
IgG
20 μL
-20°C
Primary
148870
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
CCDC27
Liquid
CCDC27
CCDC27 Fusion Protein Ag20676
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers