Learn More
CCDC6 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98985MFV610
3396.00 SEK valid until 2025-03-28
Use promo code "25306" to get your promotional price.
Product Code. 18515824
  • 4245.00 SEK / 0.10mL
Explore more special offers
Description
Description
CCDC6 Polyclonal antibody specifically detects CCDC6 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunocytochemistry
Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
coiled-coil domain containing 6, D10S170Protein H4, H4DNA segment on chromosome 10 (unique) 170, Papillary thyroid carcinoma-encoded protein, PTCFLJ32286, TPCcoiled-coil domain-containing protein 6, TST1
Purified
Polyclonal
50mM Sodium Borate
Store at 4C in the dark.