missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC71L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564350
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ESCPAKPVAP GPCFGGRTLE EIWRAAT Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 67%.
CCDC71L Polyclonal antibody specifically detects CCDC71L in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
CCDC71L | |
Polyclonal | |
Unconjugated | |
CCDC71L | |
C7orf74; CCDC71L; coiled-coil domain containing 71-like; coiled-coil domain-containing protein 71L | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
168455 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N9Z2 | |
CCDC71L | |
Recombinant protein corresponding to Human CCDC71L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only