Läs mer
CCG-63808, MedChemExpress
Produktkod. 16418198
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16418198 10 mg
16414097 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16418198 Leverantör Medchem Express Leverantörsnummer HY70075
MedChemExpress CCG-63808 is a reversible inhibitor of regulator of G-protein signaling (RGS) proteins.
CCG-63808 is a reversible inhibitor of regulator of G-protein signaling (RGS) proteins.
Specifikationer
Kemiskt namn eller material CCG-63808
CAS 620113-73-7
Färg Red
Anteckningar om renhetsgrad Research
Molekylformel C25H15FN4O2S
Kvantitet 10 mg
Löslighetsinformation DMSO : 1 mg/mL (2.20 mM; Need ultrasonic)
LEDER N#C/C(C1=NC2=CC=CC=C2S1)=C\C3=C(N=C4C(C)=CC=CN4C3=O)OC5=CC=C(C=C5)F
Molekylvikt (g/mol) 454.48
Formel vikt 454.48
Procent renhet 97.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death