missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CCL14 Recombinant Protein

Brand:  Abnova™ H00006358-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16120592

  • 4515.00 SEK / 10µg
Estimated Shipment: 02-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH45165
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
33.88
8
Glutathione Sepharose 4 Fast Flow
10 ug
TKTESSSRGPYHPSECCFTYTTYKIPRQRIMDYYETNSQCSKPGIVFITKRGHSVCTNPSDKWVQDYIKDMKEN
CC-1/CC-3/CKb1/HCC-1/HCC-3/MCIF/NCC-2/NCC2/SCYA14/SCYL2/SY14
CCL14
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
6358
CCL14 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CCL14
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers