Läs mer
CCL21/6Ckine Antibody (59106), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB457MFV610
Produktkod. 18514389
  • 6700.00 SEK / 0.10ml
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Beskrivning
Beskrivning
CCL21/6Ckine Monoclonal antibody specifically detects CCL21/6Ckine in Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair), Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifikationer
Specifikationer
CCL21/6Ckine
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Rat
0.1 mL
Biologically Active Proteins, Cancer, Cell Cycle and Replication, Chemokines and Cytokines
6366
Store at 4C in the dark.
Western Blot, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
59106
Western Blot, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, CyTOF-ready
6CkineSmall-inducible cytokine A21, Beta-chemokine exodus-2, chemokine (C-C motif) ligand 21, CKb9, member 21, SCYA21MGC34555, secondary lymphoid tissue chemokine, SLCSecondary lymphoid-tissue chemokine
E. coli-derived recombinant mouse CCL21/6Ckine, Ser24-Gly133, Accession # P84444
RUO
Primary
Mouse
IgG2b