Läs mer
PeproTech CCL23 Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: PeproTech 500-P124-1MG
Beskrivning
AA Sequence of recombinant protein: RVTKDAETEF MMSKLPLENP VLLDRFHATS ADCCISYTPR SIPCSLLESY FETNSECSKP GVIFLTKKGR RFCANPSDKQ VQVCMRMLKL DTRIKTRKN. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human MIP-3 (CCL23). Anti-Human MIP-3 (CCL23)-specific antibody was purified by affinity chromatography employing an immobilized Human MIP-3 (CCL23) matrix. Sandwich ELISA: To detect Human MIP-3 (CCL23) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human MIP-3 (CCL23) (500-P124Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human MIP-3 (CCL23). Western Blot: To detect Human MIP-3 (CCL23) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human MIP-3 (CCL23) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
This gene is one of several chemokine genes clustered on the q-arm of chromosome 17. Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of the N-terminal cysteine residues of the mature peptide. This chemokine, a member of the CC subfamily, displays chemotactic activity on resting T lymphocytes and monocytes, lower activity on neutrophils and no activity on activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line. In addition, the product of this gene is a potent agonist of the chemokine (C-C motif) receptor 1. Alternative splicing results in multiple transcript variants that encode different isoforms.
Specifikationer
| CCL23 | |
| Polyclonal | |
| Unconjugated | |
| CCL23 | |
| C6 beta-chemokine; C-C motif chemokine 23; C-C motif chemokine ligand 23; CCL23; CCL23(19-99); CCL23(22-99); CCL23(27-99); CCL23(30-99); chemokine (C-C motif) ligand 23; Ck beta 8-1; CKb8; CKB-8; Ckb-8-1; CK-BETA-8; hmrp-2a; Macrophage inflammatory protein 3; MIP3; MIP-3; MIP-3CK-BETA-8; MPIF1; MPIF-1; MPIF-1Ckb-8-1; Myeloid progenitor inhibitory factor 1; SCYA23; small inducible cytokine subfamily A (Cys-Cys), member 23; Small-inducible cytokine A23 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6368 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P55773 | |
| CCL23 | |
| E.coli-derived Recombinant Human MIP-3 (CCL23). | |
| 1 mg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.