missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCPG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13861-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Acts as an assembly platform for Rho protein signaling complexes. Limits guanine nucleotide exchange activity of MCF2L toward RHOA, which results in an inhibition of both its transcriptional activation ability and its transforming activity. Does not inhibit activity of MCF2L toward CDC42, or activity of MCF2 toward either RHOA or CDC42. May be involved in cell cycle regulation.Specifications
CCPG1 | |
Polyclonal | |
Unconjugated | |
CCPG1 | |
CCP8, CCPG1, cell cycle progression 1, CPR8, KIAA1254 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
363098, 72278, 9236 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q640L3, Q9ULG6 | |
Ccpg1 | |
CCPG1 Fusion Protein Ag4841 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |