missing translation for 'onlineSavingsMsg'
Läs mer

CCR1 Mouse anti-Human, Clone:8E10G6, Abnova™

Produktkod. 16084906
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
Förpackningsstorlek:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16084906 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16084906 Leverantör Abnova Leverantörsnummer MAB17627.100ug

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Mouse monoclonal antibody raised against recombinant human CCR1.

This gene encodes a member of the beta chemokine receptor family, which is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. The ligands of this receptor include macrophage inflammatory protein 1 alpha (MIP-1 alpha), regulated on activation normal T expressed and secreted protein (RANTES), monocyte chemoattractant protein 3 (MCP-3), and myeloid progenitor inhibitory factor-1 (MPIF-1). Chemokines and their receptors mediated signal transduction are critical for the recruitment of effector immune cells to the site of inflammation. Knockout studies of the mouse homolog suggested the roles of this gene in host protection from inflammatory response, and susceptibility to virus and parasite. This gene and other chemokine receptor genes, including CCR2, CCRL2, CCR3, CCR5 and CCXCR1, are found to form a gene cluster on chromosome 3p. [provided by RefSeq]

Specifikationer

Antigen CCR1
Användningsområden ELISA, Flow Cytometry, Western Blot
Klassificering Monoclonal
Klona 8E10G6
Konjugera Unconjugated
Beskrivning chemokine (C-C motif) receptor 1
Utspädning ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunocytochemistry
Flow Cytometry (1:200-1:400)
Immunohistochemistry
The optimal working dilution should be determined by the end user.
Formulering In PBS (0.05% sodium azide)
Gen CCR1
Gene Alias CD191, CKR-1, CKR1, CMKBR1, HM145, MIP1aR, SCYAR1
Värd art Mouse
Immunogen Recombinant protein corresponding to amino acid of human CCR1 from E. coli.
Kvantitet 100 μg
Gen-ID (Entrez) 1230
Målarter Human
Innehåll och lagring Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Produkttyp Antibody
Form Liquid
Isotyp IgG1
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.