missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CCR7 |
---|---|
Concentration | 0.73 mg/mL |
Applications | Immunohistochemistry (Paraffin), Flow Cytometry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836885
|
Proteintech
55425-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16826885
|
Proteintech
55425-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CCR7 is a member of the G protein coupled receptor family, which is a subfamily of chemokines. CCR7 was identified to be induced by the Epstein Barr virus (EBV), and is thought to be a mediator of EBV effects on B lymphocytes. CCR7 has been reported to be expressed in blood, bone marrow, lymph node, and intestine. CCR7 is particularly expressed in lymphoid tissues and in activated B and T lymphocytes and has been shown to control the migration of memory T cells to inflamed tissues, as well as stimulate dendritic cell maturation. The chemokine (C-C motif) ligand 19 (CCL19/ECL) has been reported to be a specific ligand of this receptor. ESTs have been isolated from blood, embryo, lymph node, and thymus libraries. Receptors for the C - C chemokine family include CCR 1, CCR 2A, CCR 3, CCR 4, CCR 5 and the Duffy blood group antigen. The C-C receptors are important in the function of T cell chemotaxis and migration of phagocytic cells to sites of inflammation.Specifications
CCR7 | |
Immunohistochemistry (Paraffin), Flow Cytometry | |
Unconjugated | |
Rabbit | |
Human | |
P32248 | |
1236 | |
Peptide | |
Primary | |
-20°C | |
CCR7 |
0.73 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BLR2, C C chemokine receptor type 7, C C CKR 7, CC CKR 7, CCR 7, CCR7, CD197, CDw197, CMKBR7, EBI1, EVI1, MIP 3 beta receptor | |
CCR7 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |