missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55425-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
CCR7 is a member of the G protein coupled receptor family, which is a subfamily of chemokines. CCR7 was identified to be induced by the Epstein Barr virus (EBV), and is thought to be a mediator of EBV effects on B lymphocytes. CCR7 has been reported to be expressed in blood, bone marrow, lymph node, and intestine. CCR7 is particularly expressed in lymphoid tissues and in activated B and T lymphocytes and has been shown to control the migration of memory T cells to inflamed tissues, as well as stimulate dendritic cell maturation. The chemokine (C-C motif) ligand 19 (CCL19/ECL) has been reported to be a specific ligand of this receptor. ESTs have been isolated from blood, embryo, lymph node, and thymus libraries. Receptors for the C - C chemokine family include CCR 1, CCR 2A, CCR 3, CCR 4, CCR 5 and the Duffy blood group antigen. The C-C receptors are important in the function of T cell chemotaxis and migration of phagocytic cells to sites of inflammation.Specifications
CCR7 | |
Polyclonal | |
Unconjugated | |
CCR7 | |
BLR2, C C chemokine receptor type 7, C C CKR 7, CC CKR 7, CCR 7, CCR7, CD197, CDw197, CMKBR7, EBI1, EVI1, MIP 3 beta receptor | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1236 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
0.73 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P32248 | |
CCR7 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |