Läs mer
Tocris Bioscience™ CCT 018159
GREENER_CHOICE
Produktkod. 18054622
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18054622 10 mg
18034402 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18054622 Leverantör Tocris Bioscience™ Leverantörsnummer 2435/10
Hsp90 inhibitor
Novel inhibitor of heat shock protein 90 (Hsp90) ATPase activity (IC50 = 5.7 μM) that displays selectivity over human Hsp72 and topoisomerase II. Inhibits proliferation of HCT116 human colon tumor cells and produces upregulation of Hsp70 and downregulation of c-Raf and cdk4. More soluble than 17-AAG (Cat. No. 1515) and is independent of NQO1/DT-diaphorase and P-glycoprotein expression.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 171009-07-7
Molekylformel C20H20N2O4
Specifikationer
Kemiskt namn eller material CCT 018159
Kvantitet 10 mg
Formel vikt 352.39
Renhet >98%
Molekylvikt (g/mol) 352.39