Läs mer
CCT036477, TRC
Produktkod. 30361357
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30361357 250 mg
30360413 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30361357 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC227900250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 305372-78-5
Molekylformel C21H18ClN3
Molekylvikt (g/mol) 347.84
Synonym α-(4-Chlorophenyl)-2-methyl-N-2-pyridinyl-1H-indole-3-methanamine, CCT036477
IUPAC-namn N-[(4-chlorophenyl)-(2-methyl-1H-indol-3-yl)methyl]pyridin-2-amine
LEDER Cc1[nH]c2ccccc2c1C(Nc3ccccn3)c4ccc(Cl)cc4
Specifikationer
Kemiskt namn eller material alpha-(4-Chlorophenyl)-2-methyl-N-2-pyridinyl-1H-indole-3-methanamine
Kvantitet 250 mg
InChI formel InChI=1S/C21H18ClN3/c1-14-20(17-6-2-3-7-18(17)24-14)21(15-9-11-16(22)12-10-15)25-19-8-4-5-13-23-19/h2-13,21,24H,1H3,(H,23,25)