missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCT7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15994-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
TPRBK may be involved in the condensation of spindle midzone microtubules, leading to the formation of midbody. Diseases associated with this gene include Cleft Soft Palate.Specifications
CCT7 | |
Polyclonal | |
Unconjugated | |
CCT7 | |
CCT ETA, CCT7, Ccth, HIV 1 Nef interacting protein, Nip7 1, TCP 1 eta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10574, 12468, 297406 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P80313, Q99832 | |
CCT7 | |
CCT7 Fusion Protein Ag8868 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |