Läs mer
Tocris Bioscience™ CD 2314
GREENER_CHOICE
Produktkod. 30002012
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30002012 50 mg
30001998 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30002012 Leverantör Tocris Bioscience™ Leverantörsnummer 3824/50
Selective RARβ agonist
Potent and selective RARβ receptor agonist (Kd values are 145 and >3760 nM for RARβ and RARα receptors respectively; no binding detected at RARγ). Inhibits growth of human HNSCC 22B, 183A and 22A cells (IC50 values are 3.0, 5.7 and 8.0 μM respectively).
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 170355-37-0
Molekylformel C23H24O2S
Specifikationer
Kemiskt namn eller material CD 2314
Kvantitet 50 mg
Formel vikt 364.5
Fysisk form Solid
Färg White
Renhet >98%
Molekylvikt (g/mol) 364.503