Läs mer
Tocris Bioscience™ CD 2665
GREENER_CHOICE
Produktkod. 18094492
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18094492 10 mg
18015712 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18094492 Leverantör Tocris Bioscience™ Leverantörsnummer 3800/10
Selective RARβ/γ antagonist
Selective RARβγ antagonist (KD values are 110, 306 and > 1000 nM for RARγ, RARβ and RARα respectively). Blocks retinoic acid-induced apoptosis ex vivo.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 170355-78-9
Molekylformel C31H34O5
Specifikationer
Kemiskt namn eller material CD 2665
Kvantitet 10 mg
Formel vikt 486.6
Renhet >98%
Molekylvikt (g/mol) 486.608