Läs mer
Tocris Bioscience™ CD 437
GREENER_CHOICE
Produktkod. 18712744
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18712744 10 mg
18792534 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18712744 Leverantör Tocris Bioscience™ Leverantörsnummer 1549/10
RARγ-selective agonist
Synthetic retinoid that is an RARγ-selective agonist. Displays RARγ-dependent and -independent effects on differentiation and apoptosis.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 125316-60-1
Molekylformel C27H26O3
Specifikationer
Kemiskt namn eller material CD 437
Kvantitet 10 mg
Formel vikt 398.5
Renhet >98%
Molekylvikt (g/mol) 398.502