Learn More
CD100 (SEMA4D) Monoclonal Antibody (eBio133-1C6 (133-1C6)), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1296.00 SEK - 3155.00 SEK
Specifications
Antigen | CD100 (SEMA4D) |
---|---|
Clone | eBio133-1C6 (133-1C6) |
Concentration | 0.5 mg/mL |
Applications | ELISA, Flow Cytometry, Immunoprecipitation |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15227137
|
Affymetrix eBioscience
14-1009-82 |
100 μg |
3155.00 SEK
100µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15217137
|
Affymetrix eBioscience
14-1009-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The eBio133-1C6 monoclonal antibody reacts with human CD100, a member of the class IV semaphorin family also called sema4D. CD100 promotes angiogenesis, invasive growth, proliferation, and anti-apoptosis of cancer cells in vitro. Furthermore CD100 is a costimulatory molecule in adaptive immune responses by switching off negative signalling from CD72. In addition to CD72 another ligand is Plexin-B1. It has been shown that higher expression levels of CD100 are correlated with poor survival in soft tissue sarcoma patients. Expression is found in both hematopoietic and non-hematopoietic cells.CD100 is expressed on resting T cells and NK cells and B cell. CD100 can also exist as a soluble form. Applications Reported: This eBio133-1C6 (133-1C6) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and ELISA. Applications Tested: This eBio133-1C6 (133-1C6) antibody has been tested by flow cytometric analysis of human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE.
Semaphorins are neuronal chemorepellants that direct pioneering neurons during nervous system development. CD100 (also known as semaphorin 4d), a 150 kDa homodimer, is a novel semaphorin that induces B cells to aggregate and improves their viability in vitro. CD100 modifies CD40-CD40L B cell signaling by augmenting B cell aggregation and survival and down regulating CD23 expression. CD72 is a lymphocyte receptor for the class IV semaphorin CD100, which is involved in regulating B cell signaling. CD100 is expressed on resting and PHA stimulated T cells. The protein is weakly expressed on NK cells, EBV transformed B cells, monocytes and tumor/peripheral blood T cell lines, and at higher density on activated T cells and B cells.Specifications
CD100 (SEMA4D) | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
A8; BB18; C9orf164; CD100; coll-4; GR3; M-Sema G; M-sema-G; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4D; sema J; Sema4d; Semacl2; SEMAJ; semaphorin 4D; semaphorin H; semaphorin-4D; Semaphorin-C-like 2; semaphorin-J; Semcl2 | |
Sema4d | |
Primary | |
4° C | |
Sema4d |
eBio133-1C6 (133-1C6) | |
ELISA, Flow Cytometry, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
Q92854 | |
10507 | |
IgM | |
Affinity chromatography | |
Antibody |
For Research Use Only.