missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ CD105/Endoglin/TGFβ 1/3 Receptor Rabbit Polyclonal Antibody with BSA and azide
Obtain accurate and reliable results in immunohistochemistry experiments with Epredia™ CD105/Endoglin/TGF β 1/3 Receptor, Rabbit Polyclonal Antibody.
3656.70 SEK - 6109.80 SEK
Specifications
Antigen | CD105/Endoglin/TGF beta 1/3 Receptor |
---|---|
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12653557
|
Epredia™
RB-9291-P1 |
Concentrated | 500 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12643557
|
Epredia™
RB-9291-P |
Concentrated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
CD105 or Endoglin or GP160 is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists as an O- and N-glycosylated homodimer. Up-regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation-associated endothelial cell marker. CD105 binds to TGF-β1 and β3 with high affinity but not TGF-β2.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested
Epitope: Not known
Immunogen: Recombinant protein encoding aa 26-176 of human CD105
Molecular Weight: 95 kDa (monomer) and 190 kDa (dimer)
Positive Control: Tonsil, Placenta
Cellular Localization: Membrane and cytoplasm
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
CD105/Endoglin/TGF beta 1/3 Receptor | |
Polyclonal | |
Rabbit | |
Angiogenesis | |
Recombinant protein encoding aa 26-176 of human CD105 | |
Primary |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Human | |
IgG |