missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ CD105 Monoclonal Antibody (SN6), Alexa Fluor™ 488
Mouse Monoclonal Antibody
Brand: Invitrogen™ MHCD10520
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Based on our testing, publications, and results reported from customers, the Alexa Fluor™ 488 dye provides the best fluorescein (FITC) substitute.
CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.
Specifikationer
| CD105 | |
| Monoclonal | |
| 0.1mg/mL | |
| Assay-dependent | |
| ENG | |
| AI528660; AI662476; CD105; CD105 antigen; cell surface MJ7/18 antigen; Edg; END; Endo; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); Eng; HHT1; ORW; ORW1; S-endoglin; transmembrane glycoprotein | |
| Mouse | |
| Affinity chromatography | |
| ASR | |
| Primary | |
| 1 Each | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| SN6 | |
| Alexa Fluor 488 | |
| PBS with 4mg/mL BSA and 0.1% sodium azide | |
| P17813 | |
| ENG | |
| Human CD105 (Endoglin). | |
| 500 μL | |
| Whole antibody | |
| 2022 | |
| 0.1% Sodium azide | |
| 4°C, store in dark | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering