missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD107a (LAMP-1) Monoclonal Antibody (eBio1D4B (1D4B)), APC-eFluor™ 780, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
1496.00 SEK - 3935.00 SEK
Specifications
Antigen | CD107a (LAMP-1) |
---|---|
Clone | eBio1D4B (1D4B) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16626364
|
Affymetrix eBioscience
47-1071-82 |
100 μg |
3935.00 SEK
Each |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16616364
|
Affymetrix eBioscience
47-1071-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
CD107a (LAMP-1) Monoclonal antibody specifically detects CD107a (LAMP-1) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD107a (LAMP-1) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
16783 | |
IgG2a κ | |
Affinity chromatography | |
LAMP1 |
eBio1D4B (1D4B) | |
Flow Cytometry | |
APC-eFluor 780 | |
Rat | |
Mouse | |
P11438 | |
LAMP1 | |
Primary | |
Antibody |