Läs mer
CD117/c-kit Antibody (CL1667) - Azide and BSA Free, Novus Biologicals™
Produktkod. 30287590 Handla allt Bio Techne Produkter
Change view
Kvantitet:
100 μL
Produktkod. Kvantitet
30287590 100 μL
1 options
Produktkod. 30287590 Leverantör Novus Biologicals Leverantörsnummer NBP344073
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Mouse Monoclonal Antibody
CD117/c-kit Monoclonal antibody specifically detects CD117/c-kit in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifikationer
Antigen CD117/c-kit
Användningsområden Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry
Klassificering Monoclonal
Klona CL1667
Koncentration LYOPH
Konjugera Unconjugated
Utspädning Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulering Lyophilized in 5% Trehalose.
Gene Alias CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein
Värd art Mouse
Immunogen This antibody was generated using a recombinant protein sequence of P10721, with the exact immunogen sequence remaining proprietary.
Reningsmetod Protein A purified
Kvantitet 100 μL
Regulatorisk status RUO
Forskningsdisciplin B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases
Primär eller sekundär Primary
Gen-ID (Entrez) 3815
Rekonstitution Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL.
Målarter Human
Innehåll och lagring Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles.
Form Purified
Isotyp IgG2a