Learn More
CD117 (c-Kit) Monoclonal Antibody (ACK2), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
1920.00 SEK - 5660.00 SEK
Specifications
Antigen | CD117 (c-Kit) |
---|---|
Clone | ACK2 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16606354
|
Affymetrix eBioscience
46-1172-82 |
100 μg |
5660.00 SEK
100µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16696344
|
Affymetrix eBioscience
46-1172-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
The ACK2 MAb reacts with mouse CD117, also known as c-Kit receptor, Steel factor receptor and stem cell factor receptor. A member of the tyrosine kinase receptor family, this 145 kDa molecule is expressed by a majority of hematopoietic progenitor cells characterized in the mouse bone marrow as a small subset of cells positive for Sca-1 and Thy1 (Thy1^lo) and negative for lineage markers. The interaction of the mouse c-kit receptor and steel factor promotes the proliferation and differentiation of hematopoietic progenitor cells. CD117 is also expressed by mast cells and plays a role in signaling and activation of these cells. ACK2 has been reported to be a blocking antibody.
CD117 (c-Kit) Monoclonal antibody specifically detects CD117 (c-Kit) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD117 (c-Kit) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
16590 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
KIT |
ACK2 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Rat | |
Mouse | |
P05532 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |