missing translation for 'onlineSavingsMsg'
Learn More

CD117 (c-Kit) Rat anti-Mouse, PerCP-eFluor™ 710, Clone: ACK2, eBioscience™ Invitrogen™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-1172-82

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16606354

  • 5090.00 SEK / 100µg
Estimated Shipment: 25-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The ACK2 MAb reacts with mouse CD117, also known as c-Kit receptor, Steel factor receptor and stem cell factor receptor. A member of the tyrosine kinase receptor family, this 145 kDa molecule is expressed by a majority of hematopoietic progenitor cells characterized in the mouse bone marrow as a small subset of cells positive for Sca-1 and Thy1 (Thy1^lo) and negative for lineage markers. The interaction of the mouse c-kit receptor and steel factor promotes the proliferation and differentiation of hematopoietic progenitor cells. CD117 is also expressed by mast cells and plays a role in signaling and activation of these cells. ACK2 has been reported to be a blocking antibody.

CD117 (c-Kit) Monoclonal antibody specifically detects CD117 (c-Kit) in Mouse samples. It is validated for Flow Cytometry
Specifications

Specifications

CD117 (c-Kit)
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P05532
Rat
100 μg
Primary
Mouse
Antibody
IgG2b κ
Flow Cytometry
ACK2
PerCP-eFluor 710
KIT
belly-spot; Bs; CD117; ckit; C-Kit; c-KIT gene1; C-KIT protein; c-kit proto-oncogene protein; c-kit receptor; c-kit receptor tyrosine kinase; dominant spotting; Dominant white spotting; Fdc; Gsfsco1; Gsfsco5; Gsfsow3; KIT; kit oncogene; KIT proto-oncogene receptor tyrosine kinase; KIT proto-oncogene, receptor tyrosine kinase; mast cell growth factor receptor; mast/stem cell growth factor receptor; Mast/stem cell growth factor receptor Kit; MGF; p145 c-kit; PBT; Piebald trait protein; protein kinase; proto-oncogene c-Kit; proto-oncogene tyrosine-protein kinase Kit; receptor tyrosine kinase c-kit; SCFR; SCO1; SCO5; Sl; soluble KIT variant 1; SOW3; spotted sterile male; Ssm; Steel Factor Receptor; Tr-kit; Tyrosine-protein kinase Kit; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein; W
Affinity chromatography
RUO
16590
4° C, store in dark, DO NOT FREEZE!
Liquid
Videos
SDS
Documents
Certificates
Special Offers

Special Offers