Learn More
CD117 (c-Kit) Monoclonal Antibody (2B8), Super Bright™ 645, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
4090.00 SEK
Specifications
Antigen | CD117 (c-Kit) |
---|---|
Clone | 2B8 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15855673
|
Affymetrix eBioscience
64-1171-82 |
100 μg |
4090.00 SEK
100µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The 104D2 monoclonal antibody reacts with human CD117, also known as c-Kit , Steel factor receptor and stem cell factor receptor. A member of the tyrosine kinase receptor family, this 145 kDa molecule is expressed by hematopoietic progenitor cell subsets and mast cells. The interaction of c-Kit and Steel factor promotes proliferation and differentiation of hematopoietic progenitor cells and mast cell differentiation. CD117 is also expressed by melanocytes and plays a role in signaling and activation of these cells. Applications Reported: This 104D2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 104D2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (cat. SB-4400) to minimize any non-specific polymer interactions.
CD117 (c-Kit) Monoclonal antibody specifically detects CD117 (c-Kit) in Mouse, Porcine samples. It is validated for Flow Cytometry.Specifications
CD117 (c-Kit) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P05532, Q2HWD6 | |
KIT | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
2B8 | |
Flow Cytometry | |
Super Bright 645 | |
Rat | |
Mouse, Porcine | |
16590, 396810 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only