Läs mer
CD119 (IFN gamma Receptor 1) Monoclonal Antibody (GR-20), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
31750.00 SEK - 158800.00 SEK
Specifikationer
| Antigen | CD119 (IFN gamma Receptor 1) |
|---|---|
| Klona | GR-20 |
| Koncentration | 1 mg/mL |
| Användningsområden | Flow Cytometry, Functional Assay, Neutralization |
| Klassificering | Monoclonal |
Beskrivning
The monoclonal antibody GR-20 recognizes mouse CD119 also known as IFN gamma receptor 1. CD119 heterodimerizes with IFN gamma receptor 2 (AF-1) forming the Class II cytokine receptor. The ligand IFN gamma binds IFN gamma receptor 1 subunit but signal transduction transpires via the IFN gamma receptor 2 subunit. The receptor is involved in host defense and expression is broad including B cells, NK cells, macrophages and dendritic cells as well as other non-hematopoietic cell types. The GR-20 antibody has been reported to block IFN gamma binding to CD119. Applications Reported: This GR-20 antibody has been reported for use in flow cytometric analysis and functional assays (blocking cytokine binding to the receptor). Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/ μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
CD119 (IFN gamma Receptor 1) Monoclonal antibody specifically detects CD119 (IFN gamma Receptor 1) in Mouse samples. It is validated for Flow Cytometry, Functional Assay, NeutralizationSpecifikationer
| CD119 (IFN gamma Receptor 1) | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| antiviral protein, type 2; AVP, type 2; CD119; CD119 antigen; CDw119; Ifgr; IFN-gamma receptor 1; IFN-gammaR; IFN-gamma-R1; IFN-gamma-R-alpha; Ifngr; Ifngr1; IMD27A; IMD27B; immune interferon receptor 1; INF-g receptor; interferon gamma receptor 1; Interferon gamma receptor alpha-chain; interferon-gamma receptor 1; interferon-gamma receptor alpha chain; Nktar | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| GR-20 | |
| Flow Cytometry, Functional Assay, Neutralization | |
| Functional Grade | |
| Rat | |
| Mouse | |
| P15261 | |
| 15979 | |
| Primary | |
| 4° C | |
| IFNGR1 |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.