Learn More
CD11c, PE, clone: BU15, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD11C04
Additional Details : Weight : 0.50000kg
Description
ITGAX is the integrin alpha X chain protein. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This protein combines with the beta 2 chain (ITGB2) to form a leukocyte-specific integrin referred to as inactivated-C3b (iC3b) receptor 4 (CR4). The alpha X beta 2 complex seems to overlap the properties of the alpha M beta 2 integrin in the adherence of neutrophils and monocytes to stimulated endothelium cells, and in the phagocytosis of complement coated particles.
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.Specifications
CD11c | |
Monoclonal | |
PE | |
ITGAX | |
AI449405, Cd11c, Cr4, N418, CD11 antigen-like family member C, ITGAX, integrin aX, integrin alpha-X | |
Mouse | |
Purified | |
ASR, ASR/IVD | |
3687 | |
Store at 2 - 8 °C. | |
Liquid |
Flow Cytometry | |
BU15 | |
PBS with 10% sucrose, 4mg/mL BSA and 0.1% sodium azide | |
P20702 | |
ITGAX | |
Dendritic cells of synovial fluid | |
0.5 mL | |
Primary | |
Human | |
Antibody | |
IgG1 |