missing translation for 'onlineSavingsMsg'
Learn More

CD123 Monoclonal Antibody (6H6), PerCP-Cyanine5.5, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

1548.00 SEK - 3700.00 SEK

Specifications

Antigen CD123
Clone 6H6
Concentration 5 μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15559026
View Documents Promotion Details
Affymetrix eBioscience
45-1239-41
25 Tests
1548.00 SEK
25 tests
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15569026
View Documents Promotion Details
Affymetrix eBioscience
45-1239-42
100 Tests
3700.00 SEK
100 tests
View alternative products
Save Up to 
»
Estimated Shipment: 24-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: The 6H6 monoclonal antibody reacts with human CD123, the alpha chain of the IL-3 receptor. This 60-70 kDa transmembrane protein binds to IL-3 with low affinity by itself, and when associated with CD131 (common beta chain) binds IL-3 with high affinity. CD123 is expressed by myeloid precursors, macrophages, dendritic cells, mast cells, basophils, and megakaryocytes. Applications Reported: This 6H6 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 6H6 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 695 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

CD123 Monoclonal antibody specifically detects CD123 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD123
5 μL/Test
Monoclonal
Liquid
RUO
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
CD123; CD123 antigen; CDw123; Cyrl; cytokine receptor-like protein CYRL; hIL-3Ra; Il-3 alpha subunit; IL-3 receptor alpha chain; IL-3 receptor alpha SP2 isoform; IL-3 receptor subunit alpha; IL3R; IL-3R subunit alpha; Il3ra; IL-3RA; IL-3R-alpha; IL3RAY; IL3RX; IL3RY; interleukin 3 receptor subunit alpha; interleukin 3 receptor, alpha; interleukin 3 receptor, alpha (low affinity); interleukin 3 receptor, alpha chain; interleukin-3 receptor class II alpha chain; Interleukin-3 receptor subunit alpha; Sut-1
IL3RA
Primary
4° C, store in dark, DO NOT FREEZE!
IL3RA
6H6
Flow Cytometry
PerCP-Cyanine5.5
Mouse
Human
P26951
3563
IgG1 κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.